Learn More
MSC Vermont Gage Plug Gage, 10.28 mm Diameter, Class ZZ Plus, Tool Steel
Catalog No. 5000530585
Encompass
This Class ZZ Plus Plug Gage is designed for precise 'Go Inspection' and for accurately setting various measuring devices such as micrometers and verniers. Manufactured from durable tool steel, it features a 10.28 mm diameter, an overall length of 2.0000 inches, and a tight tolerance of 0.00508 mm. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable and consistent measurements.
  • Overall length of 2.0000 inches.
  • Tolerance of 0.00508 mm.
  • Constructed from tool steel for durability.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Features a 10 micro finish.
  • Suitable for "Go Inspection."
  • Ideal for setting micrometers, verniers, and other measuring devices.
Catalog No. 50-005-30585 Supplier MSC Supplier No. 68101336
This Class ZZ Plus Plug Gage is designed for precise 'Go Inspection' and for accurately setting various measuring devices such as micrometers and verniers. Manufactured from durable tool steel, it features a 10.28 mm diameter, an overall length of 2.0000 inches, and a tight tolerance of 0.00508 mm. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable and consistent measurements.
  • Overall length of 2.0000 inches.
  • Tolerance of 0.00508 mm.
  • Constructed from tool steel for durability.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Features a 10 micro finish.
  • Suitable for "Go Inspection."
  • Ideal for setting micrometers, verniers, and other measuring devices.