Learn More
MSC Vermont Gage Plug Gage, 11.14 mm, Class ZZ Plus, Tool Steel
Catalog No. 5000527916
Encompass
This plug gage is designed for precise 'Go' inspection and for setting various measuring devices. Made from durable tool steel, it features a 11.14 mm diameter with a Class ZZ Plus tolerance, ensuring accuracy for critical measurements.
  • For go inspection
  • Used to set micrometers, verniers and other measuring devices
  • Class ZZ plus tolerance
  • Constructed from tool steel
  • Reversible style
  • 10 microfinish
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
Catalog No. 50-005-27916 Supplier MSC Supplier No. 68102193
This plug gage is designed for precise 'Go' inspection and for setting various measuring devices. Made from durable tool steel, it features a 11.14 mm diameter with a Class ZZ Plus tolerance, ensuring accuracy for critical measurements.
  • For go inspection
  • Used to set micrometers, verniers and other measuring devices
  • Class ZZ plus tolerance
  • Constructed from tool steel
  • Reversible style
  • 10 microfinish
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards