Learn More
MSC Vermont Gage Plug Gage, 11.50 mm Diameter, Class ZZ Plus, Tool Steel
Catalog No. 5000530989
Encompass
This Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection. With a diameter of 11.50 mm, it is commonly used for 'Go' inspection, setting micrometers, verniers, and other measuring devices. Manufactured from durable tool steel, it offers reliable performance.
  • Accurate 11.50 mm diameter for precise gauging
  • Features Class ZZ Plus tolerance of 0.00508 mm for high accuracy
  • Constructed from robust tool steel with Rc 60/65 hardness for durability
  • Finished with a 10 microfinish or better for smooth operation
  • Suitable for 'Go' inspection, setting various measuring instruments
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards for quality assurance
  • Overall length of 2.0000 inches
Catalog No. 50-005-30989 Supplier MSC Supplier No. 68102557
This Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection. With a diameter of 11.50 mm, it is commonly used for 'Go' inspection, setting micrometers, verniers, and other measuring devices. Manufactured from durable tool steel, it offers reliable performance.
  • Accurate 11.50 mm diameter for precise gauging
  • Features Class ZZ Plus tolerance of 0.00508 mm for high accuracy
  • Constructed from robust tool steel with Rc 60/65 hardness for durability
  • Finished with a 10 microfinish or better for smooth operation
  • Suitable for 'Go' inspection, setting various measuring instruments
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards for quality assurance
  • Overall length of 2.0000 inches