Learn More
MSC Vermont Gage Plug Gage, 11.75 mm Diameter, Class ZZ Plus, 2.0000 in Overall Length
Catalog No. 5000531895
Encompass
This plug gage is a Class ZZ Plus model with an 11.75 mm diameter and 2.0000 inch overall length. It features a tolerance of 0.00508 mm and a 10 Micro finish. Designed for accurate "Go" inspection, it is also suitable for setting micrometers, verniers, and other measuring devices. A Certificate of Accuracy with NIST Traceability is included, and the gage conforms to ASME B89.1 standards.
  • Has a 2.0000 inch overall length.
  • Features a tolerance of 0.00508 mm.
  • Constructed from tool steel.
  • Suitable for "Go" inspection and setting measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Features a 10 Micro coating/finish.
  • Measures 11.75 mm in diameter.
Catalog No. 50-005-31895 Supplier MSC Supplier No. 68102805
This plug gage is a Class ZZ Plus model with an 11.75 mm diameter and 2.0000 inch overall length. It features a tolerance of 0.00508 mm and a 10 Micro finish. Designed for accurate "Go" inspection, it is also suitable for setting micrometers, verniers, and other measuring devices. A Certificate of Accuracy with NIST Traceability is included, and the gage conforms to ASME B89.1 standards.
  • Has a 2.0000 inch overall length.
  • Features a tolerance of 0.00508 mm.
  • Constructed from tool steel.
  • Suitable for "Go" inspection and setting measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Features a 10 Micro coating/finish.
  • Measures 11.75 mm in diameter.