Learn More
MSC Vermont Gage Plug Gage, 13.29 mm Diameter, 2.0000 in Length
Catalog No. 5000527298
Encompass
This Class ZZ Plus Plug Gage features a 13.29 mm diameter and a 2.0000 in overall length. It is constructed from tool steel with a 10 Micro finish, offering a tolerance of 0.00508 mm. Designed for "Go Inspection," it is ideal for setting micrometers, verniers, and other measuring devices. This gage adheres to ASME B89.1 Standards and includes a Certificate of Accuracy with NIST Traceability.
  • Diameter of 13.29 mm.
  • Overall length of 2.0000 inches.
  • Tolerance of 0.00508 mm.
  • 10 Micro finish.
  • Constructed from tool steel.
  • Designed for "Go Inspection."
  • Suitable for setting measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Adheres to ASME B89.1 standards.
Catalog No. 50-005-27298 Supplier MSC Supplier No. 68104348
This Class ZZ Plus Plug Gage features a 13.29 mm diameter and a 2.0000 in overall length. It is constructed from tool steel with a 10 Micro finish, offering a tolerance of 0.00508 mm. Designed for "Go Inspection," it is ideal for setting micrometers, verniers, and other measuring devices. This gage adheres to ASME B89.1 Standards and includes a Certificate of Accuracy with NIST Traceability.
  • Diameter of 13.29 mm.
  • Overall length of 2.0000 inches.
  • Tolerance of 0.00508 mm.
  • 10 Micro finish.
  • Constructed from tool steel.
  • Designed for "Go Inspection."
  • Suitable for setting measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Adheres to ASME B89.1 standards.