Learn More
MSC Vermont Gage Plug Gage, 14.66 mm Diameter, Class ZZ Plus, Tool Steel
Catalog No. 5000528921
Encompass
This Class ZZ Plus Plug Gage features a 0.00508 mm tolerance and a 10 micro finish. It is ideal for go inspection, setting micrometers, verniers, and other measuring devices. Crafted from tool steel, it has an overall length of 2.0000 inches and a diameter of 14.66 mm. A Certificate of Accuracy with NIST Traceability is included, and the gage meets ASME B89.1 standards.
  • Overall length of 2.0000 inches
  • Tolerance of 0.00508 mm
  • Made from tool steel
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • 10 micro finish
  • Diameter of 14.66 mm
  • Manufactured in the United States
Catalog No. 50-005-28921 Supplier MSC Supplier No. 68105717
This Class ZZ Plus Plug Gage features a 0.00508 mm tolerance and a 10 micro finish. It is ideal for go inspection, setting micrometers, verniers, and other measuring devices. Crafted from tool steel, it has an overall length of 2.0000 inches and a diameter of 14.66 mm. A Certificate of Accuracy with NIST Traceability is included, and the gage meets ASME B89.1 standards.
  • Overall length of 2.0000 inches
  • Tolerance of 0.00508 mm
  • Made from tool steel
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • 10 micro finish
  • Diameter of 14.66 mm
  • Manufactured in the United States