Learn More
MSC Vermont Gage Plug Gage, 2.02 mm Diameter, 0.00508 mm Tolerance, Tool Steel
Catalog No. 5000529021
Encompass
This plug gage is designed for precise 'Go' inspection and calibration of various measuring devices. It features a 2.02 mm diameter with a 0.00508 mm tolerance, an overall length of 2.000 inches, and is constructed from durable tool steel with a 10 micro finish. It is suitable for setting micrometers, verniers, and other measurement equipment.
  • Provides high precision with 0.00508 mm tolerance.
  • Constructed from durable tool steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed for "Go" inspection applications.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Features a 10 micro finish.
  • Meets ASME B89.1 standards.
  • Has an overall length of 2.000 inches.
Catalog No. 50-005-29021 Supplier MSC Supplier No. 68093079
This plug gage is designed for precise 'Go' inspection and calibration of various measuring devices. It features a 2.02 mm diameter with a 0.00508 mm tolerance, an overall length of 2.000 inches, and is constructed from durable tool steel with a 10 micro finish. It is suitable for setting micrometers, verniers, and other measurement equipment.
  • Provides high precision with 0.00508 mm tolerance.
  • Constructed from durable tool steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed for "Go" inspection applications.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Features a 10 micro finish.
  • Meets ASME B89.1 standards.
  • Has an overall length of 2.000 inches.