Learn More
MSC Vermont Gage Plug Gage, 2.48 mm, Class ZZ Plus, Tool Steel
Catalog No. 5000527911
Encompass
This plug gage is an essential tool for precise measurement and inspection, designed to verify hole sizes and set various measuring devices. Constructed from durable tool steel, it offers reliability and accuracy for demanding industrial applications.
  • Constructed from tool steel with a Rockwell hardness of 60 to 65.
  • Features a 10 microfinish or better for enhanced precision.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed for go inspection and setting micrometers, verniers, and other measuring devices.
  • Overall length of 2 inches.
  • Conforms to ASME B89.1 standards.
  • Manufactured in the United States.
Catalog No. 50-005-27911 Supplier MSC Supplier No. 68093533
This plug gage is an essential tool for precise measurement and inspection, designed to verify hole sizes and set various measuring devices. Constructed from durable tool steel, it offers reliability and accuracy for demanding industrial applications.
  • Constructed from tool steel with a Rockwell hardness of 60 to 65.
  • Features a 10 microfinish or better for enhanced precision.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed for go inspection and setting micrometers, verniers, and other measuring devices.
  • Overall length of 2 inches.
  • Conforms to ASME B89.1 standards.
  • Manufactured in the United States.