Learn More
MSC Vermont Gage Plug Gage, 21.23 mm, Class ZZ Plus, Tool Steel
Catalog No. 5000530690
Encompass
This plug gage is designed for precise 'Go' inspection, as well as for setting micrometers, verniers, and other measuring devices. It features a 21.23 mm diameter, Class ZZ Plus tolerance with a 0.00508 mm tolerance, and is constructed from durable tool steel.
  • Designed for 'Go' inspection applications
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Manufactured from high-quality tool steel
  • Features a 21.23 mm diameter and Class ZZ Plus tolerance
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Reversible design
  • Metric system of measurement
Catalog No. 50-005-30690 Supplier MSC Supplier No. 68112283
This plug gage is designed for precise 'Go' inspection, as well as for setting micrometers, verniers, and other measuring devices. It features a 21.23 mm diameter, Class ZZ Plus tolerance with a 0.00508 mm tolerance, and is constructed from durable tool steel.
  • Designed for 'Go' inspection applications
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Manufactured from high-quality tool steel
  • Features a 21.23 mm diameter and Class ZZ Plus tolerance
  • Includes a certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Reversible design
  • Metric system of measurement