Learn More
MSC Vermont Gage Plug Gage, 4.30 mm, Class ZZ Plus, Steel, 2 in.
Catalog No. 5000527246
Encompass
This precision plug gage is designed for accurate go inspection and for setting micrometers, verniers, and other measuring devices. Crafted from high-quality tool steel, it features a 4.30 mm diameter, a 2-inch length, and a superior 10 microinch finish, ensuring reliable performance and NIST traceability.
  • Class ZZ Plus tolerance
  • Tool steel construction
  • 2 in. long
  • 10 microinch finish or better
  • Round within 0.0001 in. or 0.0025 mm
  • Marked with size and direction of tolerance
  • Certificate of accuracy included with NIST traceability
  • Suitable for go inspection
  • Ideal for setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-27246 Supplier MSC Supplier No. 68095355
This precision plug gage is designed for accurate go inspection and for setting micrometers, verniers, and other measuring devices. Crafted from high-quality tool steel, it features a 4.30 mm diameter, a 2-inch length, and a superior 10 microinch finish, ensuring reliable performance and NIST traceability.
  • Class ZZ Plus tolerance
  • Tool steel construction
  • 2 in. long
  • 10 microinch finish or better
  • Round within 0.0001 in. or 0.0025 mm
  • Marked with size and direction of tolerance
  • Certificate of accuracy included with NIST traceability
  • Suitable for go inspection
  • Ideal for setting micrometers, verniers, and other measuring devices