Learn More
MSC Vermont Gage Plug Gage, 5.85 mm, Class ZZ Plus, Tool Steel
Catalog No. 5000535830
Encompass
This Class ZZ Plus Plug Gage features a 5.85 mm diameter and an overall length of 2.0000 inches. Made from durable tool steel with a 10 microfinish, it is designed for precise 'Go' inspection and setting various measuring devices such as micrometers and verniers. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Designed for 'Go' inspection and setting measuring devices
  • Constructed from tool steel for durability
  • Features a 10 microfinish for precision
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Precise 0.00508 mm tolerance
Catalog No. 50-005-35830 Supplier MSC Supplier No. 68096908
This Class ZZ Plus Plug Gage features a 5.85 mm diameter and an overall length of 2.0000 inches. Made from durable tool steel with a 10 microfinish, it is designed for precise 'Go' inspection and setting various measuring devices such as micrometers and verniers. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Designed for 'Go' inspection and setting measuring devices
  • Constructed from tool steel for durability
  • Features a 10 microfinish for precision
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Precise 0.00508 mm tolerance