Learn More
MSC Vermont Gage Plug Gage, 5.95 mm Diameter, 0.00508 mm Tolerance, 2.0000 in Length
Catalog No. 5000532030
Encompass
This plug gage is ideal for precise measurement tasks, featuring a 5.95 mm diameter and a 0.00508 mm tolerance. Constructed from durable tool steel, it is suited for "go" inspection and calibrating micrometers, verniers, and other measuring tools. It also boasts a 10 micro surface finish and includes a Certificate of Accuracy.
  • Designed for "go" inspection and calibrating micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a 10 micro surface finish.
  • Includes a Certificate of Accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Has a diameter of 5.95 mm.
  • Provides a tolerance of 0.00508 mm.
  • Measures 2.0000 in overall length.
Catalog No. 50-005-32030 Supplier MSC Supplier No. 68097005
This plug gage is ideal for precise measurement tasks, featuring a 5.95 mm diameter and a 0.00508 mm tolerance. Constructed from durable tool steel, it is suited for "go" inspection and calibrating micrometers, verniers, and other measuring tools. It also boasts a 10 micro surface finish and includes a Certificate of Accuracy.
  • Designed for "go" inspection and calibrating micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a 10 micro surface finish.
  • Includes a Certificate of Accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Has a diameter of 5.95 mm.
  • Provides a tolerance of 0.00508 mm.
  • Measures 2.0000 in overall length.