Learn More
MSC Vermont Gage Plug Gage, 7.20 mm Diameter, 2.0000 in OAL, Tool Steel
Catalog No. 5000528063
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurements and inspection tasks. Ideal for verifying hole sizes, gaging slot widths, and setting various measuring devices, it offers reliable performance for a range of applications.
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 microinch finish or better
  • Meets ASME B89.1 standards
  • Offers economical general purpose gaging with added wear life
  • Provides quick and accurate measurements
  • Can be used to measure hole sizes and calculate distances between holes
  • Suitable for Go/NoGo gaging
Catalog No. 50-005-28063 Supplier MSC Supplier No. 68098250
This Class ZZ Plus Plug Gage is designed for precise measurements and inspection tasks. Ideal for verifying hole sizes, gaging slot widths, and setting various measuring devices, it offers reliable performance for a range of applications.
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 microinch finish or better
  • Meets ASME B89.1 standards
  • Offers economical general purpose gaging with added wear life
  • Provides quick and accurate measurements
  • Can be used to measure hole sizes and calculate distances between holes
  • Suitable for Go/NoGo gaging