Learn More
MSC Vermont Gage Plug Gage, 7.31 mm, Class ZZ Plus, Tool Steel
Catalog No. 5000513097
Encompass
This plug gage is designed for precise measurement and inspection tasks. It features a 7.31 mm diameter, an overall length of 2.0000 inches, and a tight tolerance of 0.00508 mm. Constructed from durable tool steel with a 10 micro finish, it ensures reliable performance in various applications.
  • Certificate of accuracy with NIST traceability.
  • Hassle-free guarantee.
  • Meets ASME B89.1 standards.
  • Made from high-quality tool steel.
  • Features a 10 micro finish.
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices.
Catalog No. 50-005-13097 Supplier MSC Supplier No. 68098367
This plug gage is designed for precise measurement and inspection tasks. It features a 7.31 mm diameter, an overall length of 2.0000 inches, and a tight tolerance of 0.00508 mm. Constructed from durable tool steel with a 10 micro finish, it ensures reliable performance in various applications.
  • Certificate of accuracy with NIST traceability.
  • Hassle-free guarantee.
  • Meets ASME B89.1 standards.
  • Made from high-quality tool steel.
  • Features a 10 micro finish.
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices.