Learn More
MSC Vermont Gage Plug Gage, Class X, 0.0320 in. Diameter, Go (Plus), Tool Steel
Catalog No. 5000511702
Encompass
This Class X Go Pin Gage is constructed from tool steel and features a precise 0.0320 in. diameter. Designed for accuracy, it can be used on the shop floor or in metrology laboratories to calibrate measuring equipment or directly gauge parts.
  • Class X tolerance
  • Go (Plus) gauge type
  • 0.0320 in. diameter
  • 2 in. overall length
  • Tool steel construction with RC 60/65 hardness
  • 4 microinch surface finish or better
  • Includes certificate of accuracy with NIST traceability
  • Meets or exceeds ISO 17025 standards
  • Suitable for calibrating measuring equipment
  • Suitable for direct part gauging
Catalog No. 50-005-11702 Supplier MSC Supplier No. 88703202
This Class X Go Pin Gage is constructed from tool steel and features a precise 0.0320 in. diameter. Designed for accuracy, it can be used on the shop floor or in metrology laboratories to calibrate measuring equipment or directly gauge parts.
  • Class X tolerance
  • Go (Plus) gauge type
  • 0.0320 in. diameter
  • 2 in. overall length
  • Tool steel construction with RC 60/65 hardness
  • 4 microinch surface finish or better
  • Includes certificate of accuracy with NIST traceability
  • Meets or exceeds ISO 17025 standards
  • Suitable for calibrating measuring equipment
  • Suitable for direct part gauging