Learn More
MSC Vermont Gage Plug Gage, Class X, 0.135 Inch Diameter, Go, Steel
Catalog No. 5000510310
Encompass
This Vermont Gage Class X Go Plug Gage, made of steel, features a 0.135-inch diameter and is engineered for precise measurement and verification.
  • Made of steel
  • Reversible style
  • Go direction of tolerance
  • Available with standard or expedited ISO 17025 accredited calibration options
  • Custom laser engraving available
Catalog No. 50-005-10310 Supplier MSC Supplier No. 88713508
This Vermont Gage Class X Go Plug Gage, made of steel, features a 0.135-inch diameter and is engineered for precise measurement and verification.
  • Made of steel
  • Reversible style
  • Go direction of tolerance
  • Available with standard or expedited ISO 17025 accredited calibration options
  • Custom laser engraving available