Learn More
MSC Vermont Gage Plug Gage, Class X, 0.1796 in dia., Go (Plus), Tool Steel
Catalog No. 5000531586
Encompass
This plug gage is designed for precise measurement applications in both shop floor and metrology laboratory environments. Constructed from tool steel with a 4 microinch finish or better, it features a Class X tolerance of 0.00004 " Each gage serves as a master for calibrating measuring equipment or directly gaging parts, and includes a Certificate of Accuracy with NIST Traceability, meeting ISO 17025 standards.
  • Class X tolerance
  • Go (Plus) type
  • Tool steel construction with RC 60/65 hardness
  • Chrome plated finish with 4 microinch finish or better
  • 0.00004 in tolerance
  • Suitable for shop floor and metrology laboratory applications
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards
Catalog No. 50-005-31586 Supplier MSC Supplier No. 87552923
This plug gage is designed for precise measurement applications in both shop floor and metrology laboratory environments. Constructed from tool steel with a 4 microinch finish or better, it features a Class X tolerance of 0.00004 " Each gage serves as a master for calibrating measuring equipment or directly gaging parts, and includes a Certificate of Accuracy with NIST Traceability, meeting ISO 17025 standards.
  • Class X tolerance
  • Go (Plus) type
  • Tool steel construction with RC 60/65 hardness
  • Chrome plated finish with 4 microinch finish or better
  • 0.00004 in tolerance
  • Suitable for shop floor and metrology laboratory applications
  • Includes certificate of accuracy with NIST traceability
  • Meets ISO 17025 standards