Learn More
MSC Vermont Gage Plug Gage, Class X, 0.2105 in. dia., Go (Plus)
Catalog No. 5000534400
Encompass
This Class X Go Pin Gauge is made from tool steel with a hardness of RC 60/65, featuring an uncoated steel construction and a surface finish of 4 microinch or better. It has a diameter of 0.2105 in and an overall length of 2 in, with a tolerance of 0.00004 in. This plug gage is designed for precise measurements and includes a Certificate of Accuracy with NIST Traceability, meeting ISO 17025 standards.
  • Made from tool steel with RC 60/65 hardness.
  • Uncoated steel construction with 4 microinch or better surface finish.
  • Diameter of 0.2105 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Go (plus) gauge type.
  • Suitable for shop floor and metrology laboratory applications.
  • Includes certificate of accuracy with NIST traceability.
  • Meets ISO 17025 standards.
Catalog No. 50-005-34400 Supplier MSC Supplier No. 88721055
This Class X Go Pin Gauge is made from tool steel with a hardness of RC 60/65, featuring an uncoated steel construction and a surface finish of 4 microinch or better. It has a diameter of 0.2105 in and an overall length of 2 in, with a tolerance of 0.00004 in. This plug gage is designed for precise measurements and includes a Certificate of Accuracy with NIST Traceability, meeting ISO 17025 standards.
  • Made from tool steel with RC 60/65 hardness.
  • Uncoated steel construction with 4 microinch or better surface finish.
  • Diameter of 0.2105 in.
  • Overall length of 2 in.
  • Tolerance of 0.00004 in.
  • Go (plus) gauge type.
  • Suitable for shop floor and metrology laboratory applications.
  • Includes certificate of accuracy with NIST traceability.
  • Meets ISO 17025 standards.