Learn More
MSC Vermont Gage Plug Gage, Class X, 0.2860 in. Diameter, 2 in. Long, Tool Steel
Catalog No. 5000510311
Encompass
This standard plug gage is suitable for use on both the shop floor and in metrology laboratories. It can be utilized as a master to calibrate measuring equipment or to directly gage parts, meeting or exceeding ISO 17025 standards.
  • Type Go (Plus)
  • Class X
  • Diameter 0.2860 in.
  • Overall length 2 in.
  • Material tool steel - Rc 60/65
  • Finish 4 microinch or better, chrome plated
  • Tolerance 0.00004 in.
  • Hardness RC 60/65
  • Includes certificate of accuracy with NIST traceability
  • Meets or exceeds ISO 17025 standards
Catalog No. 50-005-10311 Supplier MSC Supplier No. 88728605
This standard plug gage is suitable for use on both the shop floor and in metrology laboratories. It can be utilized as a master to calibrate measuring equipment or to directly gage parts, meeting or exceeding ISO 17025 standards.
  • Type Go (Plus)
  • Class X
  • Diameter 0.2860 in.
  • Overall length 2 in.
  • Material tool steel - Rc 60/65
  • Finish 4 microinch or better, chrome plated
  • Tolerance 0.00004 in.
  • Hardness RC 60/65
  • Includes certificate of accuracy with NIST traceability
  • Meets or exceeds ISO 17025 standards