Learn More
MSC Vermont Gage Plug Gage, Class X, 0.2960 in. Diameter, Go (Plus)
Catalog No. 5000510261
Encompass
This plug gage is designed for precise measurement and calibration in both shop and metrology laboratory settings. It is used as a master to calibrate measuring equipment or to gage parts directly.
  • Class X tolerance
  • Manufactured from tool steel with RC 60/65 hardness
  • Features a 4 microinch finish or better, chrome plated
  • Overall length of 2 in.
  • Provides a tolerance of 0.00004 in.
  • Go (Plus) type gage
  • Includes a certificate of accuracy with NIST traceability
  • Meets or exceeds ISO 17025 standards
Catalog No. 50-005-10261 Supplier MSC Supplier No. 88729603
This plug gage is designed for precise measurement and calibration in both shop and metrology laboratory settings. It is used as a master to calibrate measuring equipment or to gage parts directly.
  • Class X tolerance
  • Manufactured from tool steel with RC 60/65 hardness
  • Features a 4 microinch finish or better, chrome plated
  • Overall length of 2 in.
  • Provides a tolerance of 0.00004 in.
  • Go (Plus) type gage
  • Includes a certificate of accuracy with NIST traceability
  • Meets or exceeds ISO 17025 standards