Learn More
MSC Vermont Gage Plug Gage, Class X, Go, 0.2362 in, Tool Steel
Catalog No. 5000511415
Encompass
These Class X Go Plug Gages are used as masters to calibrate measuring equipment or to gage parts directly. They provide accurate, cost-effective measurement standards suitable for both shop floor and Metrology Laboratory environments.
  • 4 microinch finish or better
  • Includes certificate of accuracy
  • NIST traceable calibration available
  • Tool steel 60/65 Rc material
  • 2 in long
  • Round within 1/2 of tolerance
  • Go (plus) tolerance of +0.00004 in (for sizes up to 0.8250 in)
Catalog No. 50-005-11415 Supplier MSC Supplier No. 92165927
These Class X Go Plug Gages are used as masters to calibrate measuring equipment or to gage parts directly. They provide accurate, cost-effective measurement standards suitable for both shop floor and Metrology Laboratory environments.
  • 4 microinch finish or better
  • Includes certificate of accuracy
  • NIST traceable calibration available
  • Tool steel 60/65 Rc material
  • 2 in long
  • Round within 1/2 of tolerance
  • Go (plus) tolerance of +0.00004 in (for sizes up to 0.8250 in)