Learn More
MSC Vermont Gage Plug Gage, Class X Minus, 0.3935 in, Tool Steel
Catalog No. 5000516406
Encompass
The Vermont Gage Class X Minus Plug Gage is engineered for precise measurement in both shop floor and metrology laboratories. Constructed from durable tool steel, this gage has a 0.3935 inch diameter and a superior 4 microinch finish. It is ideal for calibrating measuring equipment or direct part gaging, and meets or exceeds ISO 17025 standards.
  • Includes certificate of accuracy with NIST traceability
  • Made from tool steel with Rc 60/65 hardness
  • Overall length of 2 inches
  • Tolerance of 0.00004 inches
  • Class X tolerance
  • No-Go (minus) gauge type
Catalog No. 50-005-16406 Supplier MSC Supplier No. 88839352
The Vermont Gage Class X Minus Plug Gage is engineered for precise measurement in both shop floor and metrology laboratories. Constructed from durable tool steel, this gage has a 0.3935 inch diameter and a superior 4 microinch finish. It is ideal for calibrating measuring equipment or direct part gaging, and meets or exceeds ISO 17025 standards.
  • Includes certificate of accuracy with NIST traceability
  • Made from tool steel with Rc 60/65 hardness
  • Overall length of 2 inches
  • Tolerance of 0.00004 inches
  • Class X tolerance
  • No-Go (minus) gauge type