Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Vermont Gage Plug Gage, Class X, No-Go, 0.4604 in, 2 in

Catalog No. 5000530173
Encompass

This plug gage is designed for precise measurement applications, featuring a 0.4604 inch diameter and a 2 inch length. It includes a 4 micro coating and a Certificate of Accuracy with NIST Traceability, making it suitable for calibrating measuring equipment or directly gaging parts.

  • Class X
  • 0.4604 inch diameter
  • 0.00004 inch tolerance
  • Tool steel material
  • Certificate of accuracy with NIST traceability included
  • Application: calibration of measuring equipment or direct part gaging
  • 4 micro coating/finish
  • 2 inch length

Catalog No. 50-005-30173 Supplier MSC Supplier No. 80348691
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This plug gage is designed for precise measurement applications, featuring a 0.4604 inch diameter and a 2 inch length. It includes a 4 micro coating and a Certificate of Accuracy with NIST Traceability, making it suitable for calibrating measuring equipment or directly gaging parts.

  • Class X
  • 0.4604 inch diameter
  • 0.00004 inch tolerance
  • Tool steel material
  • Certificate of accuracy with NIST traceability included
  • Application: calibration of measuring equipment or direct part gaging
  • 4 micro coating/finish
  • 2 inch length

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.