Learn More
MSC Vermont Gage Plug Gage, Class X Plus, 0.1258 in Diameter, 2 in Length
Catalog No. 5000513545
Encompass
This Class X Plus Plug Gage is a precise measurement standard designed for calibrating measuring equipment or directly gaging parts. Manufactured from tool steel, it offers high accuracy and includes a Certificate of Accuracy with NIST traceability.
  • Features a 4 microinch finish for reliable performance.
  • Constructed from durable tool steel for longevity.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for the calibration of measuring equipment or direct part gaging.
  • Maintains roundness within 1/2 of its tolerance.
Catalog No. 50-005-13545 Supplier MSC Supplier No. 00771311
This Class X Plus Plug Gage is a precise measurement standard designed for calibrating measuring equipment or directly gaging parts. Manufactured from tool steel, it offers high accuracy and includes a Certificate of Accuracy with NIST traceability.
  • Features a 4 microinch finish for reliable performance.
  • Constructed from durable tool steel for longevity.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for the calibration of measuring equipment or direct part gaging.
  • Maintains roundness within 1/2 of its tolerance.