Learn More
MSC Vermont Gage Plug Gage, Class ZZ, 20.67 mm Diameter, Tool Steel
Catalog No. 5000530816
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is a high-precision measuring tool designed for accurate 'Go' inspection in various industrial applications. Constructed from durable tool steel, it features a a 20.67 mm diameter and a tight 0.00508 mm tolerance. This gage is ideal for setting micrometers, verniers, and other measuring devices, ensuring precision and reliability in quality control processes. It comes with a Certificate of Accuracy with NIST Traceability, adhering to ASME B89.1 standards.
  • High-precision plug gage for 'Go' inspection.
  • Constructed from durable tool steel.
  • Features a 20.67 mm diameter.
  • Maintains a tight 0.00508 mm tolerance.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Designed with a 10 micro coating/finish.
Catalog No. 50-005-30816 Supplier MSC Supplier No. 68111723
The Vermont Gage Class ZZ Plus Plug Gage is a high-precision measuring tool designed for accurate 'Go' inspection in various industrial applications. Constructed from durable tool steel, it features a a 20.67 mm diameter and a tight 0.00508 mm tolerance. This gage is ideal for setting micrometers, verniers, and other measuring devices, ensuring precision and reliability in quality control processes. It comes with a Certificate of Accuracy with NIST Traceability, adhering to ASME B89.1 standards.
  • High-precision plug gage for 'Go' inspection.
  • Constructed from durable tool steel.
  • Features a 20.67 mm diameter.
  • Maintains a tight 0.00508 mm tolerance.
  • Ideal for setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Designed with a 10 micro coating/finish.