Learn More
MSC Vermont Gage Plug Gage, Class ZZ, 20.81 mm, Plus, Tool Steel
Catalog No. 5000528057
Encompass
This plug gage is designed for precise measurement of inside diameters. Constructed from durable tool steel, it features a 20.81 mm diameter and a tight tolerance of 0.005 mm, ensuring accurate gauging for various industrial applications. This Class ZZ gage is of the Go (Plus) type, ideal for checking minimum hole sizes.
  • Class ZZ for precise measurement
  • 10 micro coating for enhanced durability
  • 20.81 mm measurement diameter
  • 2 inch length
  • Tool steel construction
  • 0.005 mm tolerance
  • Go (plus) type
Catalog No. 50-005-28057 Supplier MSC Supplier No. 68111863
This plug gage is designed for precise measurement of inside diameters. Constructed from durable tool steel, it features a 20.81 mm diameter and a tight tolerance of 0.005 mm, ensuring accurate gauging for various industrial applications. This Class ZZ gage is of the Go (Plus) type, ideal for checking minimum hole sizes.
  • Class ZZ for precise measurement
  • 10 micro coating for enhanced durability
  • 20.81 mm measurement diameter
  • 2 inch length
  • Tool steel construction
  • 0.005 mm tolerance
  • Go (plus) type