Learn More
MSC Vermont Gage Plug Gage, Class ZZ Minus, 0.3100 inch Diameter, Steel
Catalog No. 5000530712
Encompass
This plug gage is designed for precise measurement applications. It features a 0.3100 inch diameter and an overall length of 2 inches. Made from steel, it offers a tolerance of 0.000200 and a 10 micro coating/finish.
  • Set micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-30712 Supplier MSC Supplier No. 89031009
This plug gage is designed for precise measurement applications. It features a 0.3100 inch diameter and an overall length of 2 inches. Made from steel, it offers a tolerance of 0.000200 and a 10 micro coating/finish.
  • Set micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection
  • Includes certificate of accuracy with NIST traceability