Learn More
MSC Vermont Gage Plug Gage, Class ZZ Minus, 0.5750 in
Catalog No. 5000536866
Encompass
This precision plug gage is designed for accurately checking hole diameters and other dimensional measurements. Made from durable steel, it offers reliable performance for various inspection and calibration tasks. Each gage comes with a Certificate of Accuracy with NIST Traceability, ensuring high precision for critical applications.
  • 0.5750 in diameter
  • 2 in overall length
  • 0.000200 in tolerance
  • Constructed from steel
  • 10 micro coating/finish
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Country of origin: United States
Catalog No. 50-005-36866 Supplier MSC Supplier No. 89057509
This precision plug gage is designed for accurately checking hole diameters and other dimensional measurements. Made from durable steel, it offers reliable performance for various inspection and calibration tasks. Each gage comes with a Certificate of Accuracy with NIST Traceability, ensuring high precision for critical applications.
  • 0.5750 in diameter
  • 2 in overall length
  • 0.000200 in tolerance
  • Constructed from steel
  • 10 micro coating/finish
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Country of origin: United States