Learn More
MSC Vermont Gage Plug Gage, Class ZZ Minus, 5.05 mm, Steel
Catalog No. 5000529106
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. Made from durable steel, it features a 5.05 mm diameter and an overall length of 2 inches, making it suitable for various inspection tasks.
  • Diameter of measurement is 5.05 mm
  • Overall length is 2 inches
  • Tolerance of 0.00508 mm
  • Ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Constructed from steel
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-29106 Supplier MSC Supplier No. 40432502
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. Made from durable steel, it features a 5.05 mm diameter and an overall length of 2 inches, making it suitable for various inspection tasks.
  • Diameter of measurement is 5.05 mm
  • Overall length is 2 inches
  • Tolerance of 0.00508 mm
  • Ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Constructed from steel
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability