Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.0080 in Diameter, 2.0000 in Length
Catalog No. 5000516945
Encompass
This Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. With a diameter of 0.0080 inches and an overall length of 2.0000 inches, it is ideal for go inspection, setting micrometers, verniers, and other measuring devices. Manufactured from tool steel, it ensures durability and accuracy for various laboratory and industrial applications.
  • Manufactured from durable tool steel for longevity.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Features a 10 micro coating/finish.
  • Provides a precise tolerance of 0.000200 "
Catalog No. 50-005-16945 Supplier MSC Supplier No. 08475519
This Vermont Gage Class ZZ Plus Plug Gage is designed for precise measurement and inspection tasks. With a diameter of 0.0080 inches and an overall length of 2.0000 inches, it is ideal for go inspection, setting micrometers, verniers, and other measuring devices. Manufactured from tool steel, it ensures durability and accuracy for various laboratory and industrial applications.
  • Manufactured from durable tool steel for longevity.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Features a 10 micro coating/finish.
  • Provides a precise tolerance of 0.000200 "