Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.0125 in Diameter, Tool Steel
Catalog No. 5000516237
Encompass
This Class ZZ Plus Plug Gage features a 0.0125 in diameter and a 2.0000 in overall length. It is made from tool steel and has a tolerance of 0.000200 in and a 10 micro finish.
  • For Go inspection
  • Set micrometers
  • Verniers and other measuring devices
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-16237 Supplier MSC Supplier No. 88901251
This Class ZZ Plus Plug Gage features a 0.0125 in diameter and a 2.0000 in overall length. It is made from tool steel and has a tolerance of 0.000200 in and a 10 micro finish.
  • For Go inspection
  • Set micrometers
  • Verniers and other measuring devices
  • Includes certificate of accuracy with NIST traceability