Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.0290 in Diameter, Tool Steel
Catalog No. 5000528596
Encompass
This plug gage is engineered for precise "Go" inspection and calibration of various measuring instruments. Constructed from durable tool steel with a black oxide finish, it offers reliable performance and includes a Certificate of Accuracy with NIST traceability, ensuring compliance with ASME B89.1 standards.
  • Designed for precision "Go" inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a black oxide finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Adheres to ASME B89.1 standards.
  • Diameter of 0.0290 in.
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in.
Catalog No. 50-005-28596 Supplier MSC Supplier No. 50734151
This plug gage is engineered for precise "Go" inspection and calibration of various measuring instruments. Constructed from durable tool steel with a black oxide finish, it offers reliable performance and includes a Certificate of Accuracy with NIST traceability, ensuring compliance with ASME B89.1 standards.
  • Designed for precision "Go" inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a black oxide finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Adheres to ASME B89.1 standards.
  • Diameter of 0.0290 in.
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in.