Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.0560 in, Tool Steel
Catalog No. 5000535746
Encompass
This Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate go/no-go inspection, and for setting micrometers, verniers, and other measuring devices. Manufactured from durable tool steel with a 10 micro finish, it ensures high accuracy and reliability in various industrial and laboratory applications. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring compliance with quality standards.
  • Class ZZ Plus tolerance for high precision.
  • 0.0560 inch diameter for specific measurement requirements.
  • 2.0000 inch overall length for versatile use.
  • Constructed from durable tool steel for extended life.
  • Features a 10 micro finish for smooth operation and accuracy.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Suitable for go inspection and setting various measuring devices.
Catalog No. 50-005-35746 Supplier MSC Supplier No. 88905609
This Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate go/no-go inspection, and for setting micrometers, verniers, and other measuring devices. Manufactured from durable tool steel with a 10 micro finish, it ensures high accuracy and reliability in various industrial and laboratory applications. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring compliance with quality standards.
  • Class ZZ Plus tolerance for high precision.
  • 0.0560 inch diameter for specific measurement requirements.
  • 2.0000 inch overall length for versatile use.
  • Constructed from durable tool steel for extended life.
  • Features a 10 micro finish for smooth operation and accuracy.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Suitable for go inspection and setting various measuring devices.