Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.0595 in Diameter, 2.0000 in Overall Length
Catalog No. 5000516241
Encompass
This plug gage is a Class ZZ Plus model, designed for precise measurements. It features a diameter of 0.0595 in and an overall length of 2.0000 in, with a tolerance of 0.000200 in. Made from tool steel, it is suitable for various inspection and calibration tasks.
  • Class ZZ Plus for precise measurements.
  • Diameter of 0.0595 in.
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in.
  • Coating/finish of 10 micro.
  • Made of tool steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Ideal for Go inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
Catalog No. 50-005-16241 Supplier MSC Supplier No. 88905955
This plug gage is a Class ZZ Plus model, designed for precise measurements. It features a diameter of 0.0595 in and an overall length of 2.0000 in, with a tolerance of 0.000200 in. Made from tool steel, it is suitable for various inspection and calibration tasks.
  • Class ZZ Plus for precise measurements.
  • Diameter of 0.0595 in.
  • Overall length of 2.0000 in.
  • Tolerance of 0.000200 in.
  • Coating/finish of 10 micro.
  • Made of tool steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Meets ASME B89.1 standards.
  • Ideal for Go inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.