Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.0675 in Diameter, 2.0000 in Length, Tool Steel
Catalog No. 5000528565
Encompass
This precision plug gage is designed for accurate 'Go' inspection and for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel with a black oxide finish, it ensures reliable performance and longevity in various industrial applications.
  • Class ZZ Plus tolerance
  • 0.0675 inch diameter
  • 2.0000 inch overall length
  • Tolerance of 0.000200 inch
  • Made from tool steel
  • Features a black oxide coating
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Manufactured in the United States
Catalog No. 50-005-28565 Supplier MSC Supplier No. 50760677
This precision plug gage is designed for accurate 'Go' inspection and for setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel with a black oxide finish, it ensures reliable performance and longevity in various industrial applications.
  • Class ZZ Plus tolerance
  • 0.0675 inch diameter
  • 2.0000 inch overall length
  • Tolerance of 0.000200 inch
  • Made from tool steel
  • Features a black oxide coating
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Manufactured in the United States