Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.0735 in, 2.0000 in OAL
Catalog No. 5000529031
Encompass
This Class ZZ Plus Plug Gage is engineered for precise Go Inspection and for calibrating various measuring devices such as micrometers and verniers. It comes with a Certificate of Accuracy, providing NIST Traceability, and meets ASME B89.1 standards. The gage features a 10 Micro coating/finish for enhanced performance.
  • Designed for go inspection and setting measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Features a 10 micro coating/finish
Catalog No. 50-005-29031 Supplier MSC Supplier No. 88907357
This Class ZZ Plus Plug Gage is engineered for precise Go Inspection and for calibrating various measuring devices such as micrometers and verniers. It comes with a Certificate of Accuracy, providing NIST Traceability, and meets ASME B89.1 standards. The gage features a 10 Micro coating/finish for enhanced performance.
  • Designed for go inspection and setting measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Features a 10 micro coating/finish