Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.0870 in, Tool Steel
Catalog No. 5000530741
Encompass
This Class ZZ Plus plug gage is designed for precise 'Go' inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.0870 inch diameter and a 2.0000 inch overall length. Constructed from durable tool steel with a 10 micro finish, it meets ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for 'Go' inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a 10 micro finish.
  • Conforms to ASME B89.1 standards.
Catalog No. 50-005-30741 Supplier MSC Supplier No. 88908702
This Class ZZ Plus plug gage is designed for precise 'Go' inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.0870 inch diameter and a 2.0000 inch overall length. Constructed from durable tool steel with a 10 micro finish, it meets ASME B89.1 standards and includes a Certificate of Accuracy with NIST Traceability.
  • Includes certificate of accuracy with NIST traceability.
  • Suitable for 'Go' inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Constructed from durable tool steel.
  • Features a 10 micro finish.
  • Conforms to ASME B89.1 standards.