Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.0960 in, Tool Steel, Black Oxide
Catalog No. 5000528196
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and calibration in various applications. It provides reliable performance for quality control and instrument calibration.
  • Accurately measures 0.0960 inch diameter.
  • Features a tight tolerance of 0.000200 "
  • Constructed from durable tool steel with a black oxide finish.
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Offers an overall length of 2.0000 inches.
Catalog No. 50-005-28196 Supplier MSC Supplier No. 32911224
This Class ZZ Plus plug gage is designed for precise measurement and calibration in various applications. It provides reliable performance for quality control and instrument calibration.
  • Accurately measures 0.0960 inch diameter.
  • Features a tight tolerance of 0.000200 "
  • Constructed from durable tool steel with a black oxide finish.
  • Ideal for go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
  • Offers an overall length of 2.0000 inches.