Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.0990 in Dia, 2.0000 in OAL
Catalog No. 5000528624
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications. With a diameter of 0.0990 inch and an overall length of 2.0000 inch, it is constructed from tool steel with a black oxide finish. It is ideal for go inspection and for setting micrometers, verniers, and other measuring devices.
  • Provides accurate measurements for go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured from durable tool steel
  • Features a black oxide finish for enhanced durability
  • Meets ASME B89.1 standards
Catalog No. 50-005-28624 Supplier MSC Supplier No. 53424644
This Class ZZ Plus plug gage is designed for precise measurement applications. With a diameter of 0.0990 inch and an overall length of 2.0000 inch, it is constructed from tool steel with a black oxide finish. It is ideal for go inspection and for setting micrometers, verniers, and other measuring devices.
  • Provides accurate measurements for go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured from durable tool steel
  • Features a black oxide finish for enhanced durability
  • Meets ASME B89.1 standards