Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.1055 inch Dia, Tool Steel
Catalog No. 5000528856
Encompass
This plug gage is a Class ZZ Plus type with a 0.1055 inch diameter, manufactured from durable tool steel. It is designed for precise 'Go Inspection' and for accurately setting various measuring devices such as micrometers and verniers. The gage adheres to ASME B89.1 Standards and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements.
  • Designed for 'Go Inspection' applications.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Adheres to ASME B89.1 standards for quality assurance.
  • Features a tolerance of 0.000200 "
  • Has a 10 Micro finish for precise measurements.
Catalog No. 50-005-28856 Supplier MSC Supplier No. 88910559
This plug gage is a Class ZZ Plus type with a 0.1055 inch diameter, manufactured from durable tool steel. It is designed for precise 'Go Inspection' and for accurately setting various measuring devices such as micrometers and verniers. The gage adheres to ASME B89.1 Standards and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements.
  • Designed for 'Go Inspection' applications.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Adheres to ASME B89.1 standards for quality assurance.
  • Features a tolerance of 0.000200 "
  • Has a 10 Micro finish for precise measurements.