Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.1105 in, Tool Steel
Catalog No. 5000511321
Encompass
This tool steel Class ZZ Plus plug gage features a 0.1105 inch diameter and a 2.0000 inch overall length. It is designed for precise measurement applications, including go inspection and the setting of various measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Used for go inspection.
  • Sets micrometers, verniers, and other measuring devices.
  • Features a tolerance of 0.000200 "
  • Coating/finish is 10 micro.
  • Conforms to ASME B89.1 standards.
Catalog No. 50-005-11321 Supplier MSC Supplier No. 88911052
This tool steel Class ZZ Plus plug gage features a 0.1105 inch diameter and a 2.0000 inch overall length. It is designed for precise measurement applications, including go inspection and the setting of various measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Used for go inspection.
  • Sets micrometers, verniers, and other measuring devices.
  • Features a tolerance of 0.000200 "
  • Coating/finish is 10 micro.
  • Conforms to ASME B89.1 standards.