Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.1245 in Diameter, Tool Steel
Catalog No. 5000536739
Encompass
This Class ZZ Plus Plug Gage is engineered for precision measurement and inspection tasks. Constructed from durable tool steel, it features a 0.1245 in diameter and a 2.0000 in overall length. With a tight tolerance of 0.000200 in, this gage is ideal for 'Go' inspection and for accurately setting micrometers, verniers, and other measuring devices. It comes with a Certificate of Accuracy with NIST Traceability, ensuring compliance with industry standards such as ASME B89.1.
  • Constructed from tool steel
  • 0.1245 in diameter of measurement
  • 2.0000 in overall length
  • 0.000200 in tolerance
  • 10 micro coating/finish
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
Catalog No. 50-005-36739 Supplier MSC Supplier No. 88912456
This Class ZZ Plus Plug Gage is engineered for precision measurement and inspection tasks. Constructed from durable tool steel, it features a 0.1245 in diameter and a 2.0000 in overall length. With a tight tolerance of 0.000200 in, this gage is ideal for 'Go' inspection and for accurately setting micrometers, verniers, and other measuring devices. It comes with a Certificate of Accuracy with NIST Traceability, ensuring compliance with industry standards such as ASME B89.1.
  • Constructed from tool steel
  • 0.1245 in diameter of measurement
  • 2.0000 in overall length
  • 0.000200 in tolerance
  • 10 micro coating/finish
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards