Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.1335 in Diameter, 2.0000 in Overall Length
Catalog No. 5000530505
Encompass
This plug gage is designed for precise measurements and calibration. It features a 0.1335 inch diameter and a 2.0000 inch overall length, suitable for various inspection and calibration tasks.
  • 0.000200 inch tolerance
  • 10 micro coating/finish
  • Made from tool steel
  • Used for Go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
Catalog No. 50-005-30505 Supplier MSC Supplier No. 88913355
This plug gage is designed for precise measurements and calibration. It features a 0.1335 inch diameter and a 2.0000 inch overall length, suitable for various inspection and calibration tasks.
  • 0.000200 inch tolerance
  • 10 micro coating/finish
  • Made from tool steel
  • Used for Go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards