Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.1705 inch Diameter, Tool Steel
Catalog No. 5000532144
Encompass
This plug gage is specifically designed for precise measurement and inspection, including "Go" inspections, and for accurately setting micrometers, verniers, and other measuring devices. Crafted from durable tool steel, it features a 0.1705 inch diameter and a smooth 10 micro-finish for high accuracy. It complies with ASME B89.1 standards and includes a Certificate of Accuracy with NIST traceability.
  • Class ZZ Plus for reliable performance
  • Precisely manufactured with a 0.1705 inch diameter
  • Overall length of 2.0000 inches
  • Made from robust tool steel for durability
  • Features a smooth 10 micro-finish
  • Complies with ASME B89.1 standards
  • Includes a Certificate of Accuracy with NIST traceability
  • Ideal for Go inspection applications
  • Suitable for setting various measuring instruments
Catalog No. 50-005-32144 Supplier MSC Supplier No. 88917059
This plug gage is specifically designed for precise measurement and inspection, including "Go" inspections, and for accurately setting micrometers, verniers, and other measuring devices. Crafted from durable tool steel, it features a 0.1705 inch diameter and a smooth 10 micro-finish for high accuracy. It complies with ASME B89.1 standards and includes a Certificate of Accuracy with NIST traceability.
  • Class ZZ Plus for reliable performance
  • Precisely manufactured with a 0.1705 inch diameter
  • Overall length of 2.0000 inches
  • Made from robust tool steel for durability
  • Features a smooth 10 micro-finish
  • Complies with ASME B89.1 standards
  • Includes a Certificate of Accuracy with NIST traceability
  • Ideal for Go inspection applications
  • Suitable for setting various measuring instruments