Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.1760 in Dia, 2.0000 in OAL
Catalog No. 5000510092
Encompass
The Class ZZ Plus Plug Gage is designed for go inspection and for setting micrometers, verniers, and other measuring devices. It comes with a certificate of accuracy with NIST traceability.
  • Designed for go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Constructed from tool steel
  • Meets ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
Catalog No. 50-005-10092 Supplier MSC Supplier No. 88917604
The Class ZZ Plus Plug Gage is designed for go inspection and for setting micrometers, verniers, and other measuring devices. It comes with a certificate of accuracy with NIST traceability.
  • Designed for go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Constructed from tool steel
  • Meets ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish