Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.1770 in, 2.0000 in OAL
Catalog No. 5000535482
Encompass
This Class ZZ Plus Plug Gage features a 0.1770 in diameter and a 2.0000 in overall length. It is designed for "Go" inspection and can be used to set micrometers, verniers, and other measuring devices.
  • Class ZZ Plus plug gage
  • Made from tool steel
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Country of origin: United States
Catalog No. 50-005-35482 Supplier MSC Supplier No. 88917703
This Class ZZ Plus Plug Gage features a 0.1770 in diameter and a 2.0000 in overall length. It is designed for "Go" inspection and can be used to set micrometers, verniers, and other measuring devices.
  • Class ZZ Plus plug gage
  • Made from tool steel
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Country of origin: United States