Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.1920 in Diameter, 2.0000 in OAL
Catalog No. 5000530832
Encompass
This Class ZZ Plus Plug Gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. Made from tool steel with a black oxide finish, it features a 0.1920 inch diameter and a 2.0000 inch overall length, meeting ASME B89.1 standards. It has a 0.000200 inch tolerance and 60-62 Rockwell C hardness, and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for go inspection, setting micrometers, verniers, and other measuring devices.
  • Constructed from tool steel.
  • Black oxide coating.
  • 0.1920 inch diameter.
  • 2.0000 inch overall length.
  • 0.000200 inch tolerance.
  • 60-62 Rockwell C hardness.
  • Meets ASME B89.1 standards.
  • Includes certificate of accuracy with NIST traceability.
Catalog No. 50-005-30832 Supplier MSC Supplier No. 79090106
This Class ZZ Plus Plug Gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. Made from tool steel with a black oxide finish, it features a 0.1920 inch diameter and a 2.0000 inch overall length, meeting ASME B89.1 standards. It has a 0.000200 inch tolerance and 60-62 Rockwell C hardness, and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for go inspection, setting micrometers, verniers, and other measuring devices.
  • Constructed from tool steel.
  • Black oxide coating.
  • 0.1920 inch diameter.
  • 2.0000 inch overall length.
  • 0.000200 inch tolerance.
  • 60-62 Rockwell C hardness.
  • Meets ASME B89.1 standards.
  • Includes certificate of accuracy with NIST traceability.