Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.2065 inch, 10 microinch Finish
Catalog No. 5000528194
Encompass
This plug gage is an economical, general-purpose tool for precise measurements in various applications. Made from durable tool steel, it features a fine 10 microinch finish and Class ZZ tolerance, ensuring accuracy for critical inspections. It is ideal for measuring hole sizes, checking dimensions, and performing Go/NoGo gaging in a professional setting. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Made from tool steel with 60/65 Rc hardness
  • Features a 10 microinch finish or better
  • Designed for Go inspection applications
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Overall length is 2.0000 inch
  • Tolerance is 0.000200 inch (Class ZZ)
  • Diameter of measurement is 0.2065 inch
  • Economical for general purpose gaging
  • Can be used to measure hole sizes and distances between holes
Catalog No. 50-005-28194 Supplier MSC Supplier No. 50760370
This plug gage is an economical, general-purpose tool for precise measurements in various applications. Made from durable tool steel, it features a fine 10 microinch finish and Class ZZ tolerance, ensuring accuracy for critical inspections. It is ideal for measuring hole sizes, checking dimensions, and performing Go/NoGo gaging in a professional setting. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Made from tool steel with 60/65 Rc hardness
  • Features a 10 microinch finish or better
  • Designed for Go inspection applications
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Overall length is 2.0000 inch
  • Tolerance is 0.000200 inch (Class ZZ)
  • Diameter of measurement is 0.2065 inch
  • Economical for general purpose gaging
  • Can be used to measure hole sizes and distances between holes