Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.2085 in, 10 Microfinish
Catalog No. 5000515569
Encompass
This Class ZZ Plus plug gage features a 0.2085 inch diameter and a 2.0000 inch overall length, with a 0.000200 inch tolerance. It is constructed from tool steel and has a 10 microfinish. Designed for go inspection applications, this gage includes a Certificate of Accuracy with NIST Traceability and conforms to ASME B89.1 standards.
  • Constructed from tool steel
  • Designed for go inspection applications
  • Includes certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
  • Features a 10 microfinish
  • Overall length of 2.0000 inches
  • Tolerance of 0.000200 inches
  • Measures with a 0.2085 inch diameter
Catalog No. 50-005-15569 Supplier MSC Supplier No. 88920855
This Class ZZ Plus plug gage features a 0.2085 inch diameter and a 2.0000 inch overall length, with a 0.000200 inch tolerance. It is constructed from tool steel and has a 10 microfinish. Designed for go inspection applications, this gage includes a Certificate of Accuracy with NIST Traceability and conforms to ASME B89.1 standards.
  • Constructed from tool steel
  • Designed for go inspection applications
  • Includes certificate of accuracy with NIST traceability
  • Conforms to ASME B89.1 standards
  • Features a 10 microfinish
  • Overall length of 2.0000 inches
  • Tolerance of 0.000200 inches
  • Measures with a 0.2085 inch diameter