Learn More
MSC Vermont Gage Plug Gage, Class ZZ Plus, 0.2200 in Diameter, 2.0000 in OAL
Catalog No. 5000533133
Encompass
This Vermont Gage Class ZZ Plus Plug Gage is designed for accurate go inspection and setting various measuring devices. Constructed from durable tool steel with a black oxide finish, it provides reliable performance. Each gage includes a Certificate of Accuracy with NIST Traceability to ensure precision.
  • Designed for go inspection applications
  • Can be used to set micrometers, verniers, and other measuring devices
  • Constructed from high-quality tool steel
  • Includes a Certificate of Accuracy with NIST Traceability
  • Features a black oxide finish for enhanced durability
  • Meets ASME B89.1 standards
Catalog No. 50-005-33133 Supplier MSC Supplier No. 50743731
This Vermont Gage Class ZZ Plus Plug Gage is designed for accurate go inspection and setting various measuring devices. Constructed from durable tool steel with a black oxide finish, it provides reliable performance. Each gage includes a Certificate of Accuracy with NIST Traceability to ensure precision.
  • Designed for go inspection applications
  • Can be used to set micrometers, verniers, and other measuring devices
  • Constructed from high-quality tool steel
  • Includes a Certificate of Accuracy with NIST Traceability
  • Features a black oxide finish for enhanced durability
  • Meets ASME B89.1 standards